Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
CertifiedNunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
LidWith Lid
Array8 x 12
Packaging10 Pk., 160 Case
Product LineNunc
ColorClear
Product TypeMicroplate
MaterialPolystyrene
Volume (Metric) Well300 μL
No. of Wells96
SterilitySterile
Surface TreatmentNunclon Delta
Well ShapeRound Bottom (U-well)
No. per Case160
No. per Pack10
Unit SizeCase of 160