Goat Serum, New Zealand origin
Goat Serum, New Zealand origin
Gibco™

Goat Serum, New Zealand origin

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers withRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
16210064100 mL
16210072500 mL
Catalog number 16210064
Price (USD)/ 100 mL
69.65
Online exclusive
73.75 
Save 4.10 (6%)
-
Add to cart
Quantity:
100 mL
Price (USD)/ 100 mL
69.65
Online exclusive
73.75 
Save 4.10 (6%)
Add to cart
Goat Serum, New Zealand origin
Catalog number16210064
Price (USD)/ 100 mL
69.65
Online exclusive
73.75 
Save 4.10 (6%)
-
Add to cart
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

Sera Category: Specialty
Origin: New Zealand.
For In Vitro Diagnostic Use. For Research Use Only. Not for use in diagnostic procedures.
Specifications
AgeAdult
SpeciesGoat/Caprine
Country of OriginNew Zealand
FormLiquid
Product TypeGoat Serum
Quantity100 mL
Serum TreatmentStandard (Sterile-filtered)
SterilitySterile
Green FeaturesSustainable packaging
Product LineGibco™
Shipping ConditionFrozen
Unit Size100 mL
Contents & Storage
Storage conditions: ≤-10°C
Shipping conditions: Frozen