Pierce™ Firefly Luc One-Step Glow Assay Kit
Pierce™ Firefly Luc One-Step Glow Assay Kit
Thermo Scientific™

Pierce™ Firefly Luc One-Step Glow Assay Kit

The Thermo Scientific Pierce Firefly One-Step Luc Glow Assay Kit measures firefly luciferase activity in mammalian cell culture using aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitySufficient For
16196100 Reaction Kit100 Luciferase Assays in Microplate Wells
161971000 Reaction Kit1000 Luciferase Assays in Microplate Wells
Catalog number 16196
Price (USD)/ 100 reactions
155.65
Online exclusive
169.00 
Save 13.35 (8%)
-
Add to cart
Quantity:
100 Reaction Kit
Sufficient For:
100 Luciferase Assays in Microplate Wells
Price (USD)/ 100 reactions
155.65
Online exclusive
169.00 
Save 13.35 (8%)
Add to cart
Pierce™ Firefly Luc One-Step Glow Assay Kit
Catalog number16196
Price (USD)/ 100 reactions
155.65
Online exclusive
169.00 
Save 13.35 (8%)
-
Add to cart
The Thermo Scientific Pierce Firefly One-Step Luc Glow Assay Kit measures firefly luciferase activity in mammalian cell culture using a single reagent-addition step, making it ideal for high-throughput screening (HTS) applications.

Features of the Firefly One-Step Luc Glow Assay Kit:

Sensitive—highly sensitive detection of firefly luciferase activity
Odorless—reagents have no foul or toxic odor, eliminating the need to work in a fume hood
Convenient—one-step, mix-and-read protocol; no additional reagents required
Stable—increased signal half-life and stability compared to flash assays
Simple—does not require a luminometer with injectors
Automation-friendly—amenable to high throughput assays

This glow-type luciferase assay kit is designed for use with cultured mammalian cells that are transfected with vectors expressing the appropriate firefly luciferase reporter enzyme. Several features make the kit especially suitable for luminometers without injectors and for high-throughput screening (HTS) applications. The convenient, one-step, homogenous protocol minimizes handling steps to support the use of automation. Simply combine the supplied substrate solution and assay buffer to make a single working solution, and then add it to microplate wells containing the transfected, treated cells. The stable signal output (glow) of this system provides flexibility with regard to incubation time. When used with Thermo Scientific Firefly Luc Plasmids, the kit provides a highly sensitive bioluminescent reporter assay system for the detection of promoter or pathway activity.

Includes:
Assay buffer and substrate solution (100X)

Requires:
Firefly luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers.

Applications:
• Promoter studies for analyzing cis- and trans-acting regulatory elements
• Drug screening
• siRNA and miRNA screening
• Multiplexed assays to study off-target effects
• Protein localization reporter assays
• Signal transduction pathway analysis

Firefly luciferase is a 62kDa protein produced in nature by several species of the Lampyridae family of beetles, which includes the genera Photinus and Luciola. Thermo Scientific Red Firefly Luciferase has an emission range of 560-700nm (λmax = 613nm). Bioluminescent signal from firefly luciferase originates from the oxidation of D-Luciferin. Light output captured using a luminometer can be correlated with the amount of firefly luciferase protein produced and used to determine the activity of the promoter driving firefly luciferase expression. Firefly luciferase bioluminescence provides researchers with a non-radioactive assay for measuring transcriptional activity of regulatory elements. Firefly luciferase reporter activity can be used to determine promoter activity, to study upstream pathway activity in the cell, or to determine effect of activators or inhibitors.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypeAssay Kit
TargetLuciferase, Firefly Luciferase, Cypridina Luciferase
Sufficient For100 Luciferase Assays in Microplate Wells
Quantity100 Reaction Kit
AssayReporter Enzyme, Luciferase Reporter Assay
Compatible CellsMammalian Cells
Detection MethodBioluminescence
For Use With (Equipment)Luminometer (Microplate)
Format384-well plate, 96-well plate
Label TypeEnzyme Labeled
Substrate PropertiesChemical Substrate
TechniqueEnhanced Chemiluminescence
Substrate TypeLuciferase Substrate
SubstrateD-Luciferin
Unit Size100 reactions
Contents & Storage
• Firefly Luc One-Step Substrate, Lyophilized (makes 1μL of 1X), store at -20°C
• Firefly Luc One-Step Assay Buffer, 11 mL, store at -20°C
Upon receipt store kit at -20°C.