Specifications
Description1500 Series Class II, Type B2 BSC with UV, 6 ft (1.8m) nominal width, 230 VAC, 50/60 Hz with Type M (India) power cord and internal receptacle
Certifications/ComplianceNSF Class II Type B2 BSC
Width (Metric) Interior1841 mm
Product TypeBiosafety Cabinet
Model1500 Series
Noise Level66 dBA
Power Consumption370 W
Receptacles2 Single
Width (English) Exterior78 in.
Volume (Metric) Exhaust1696 m3/hr. (with DIM), 2149 m3/hr. (with Traverse)
Width (Metric) Exterior1981 mm
Height (Metric)1844 mm (Exterior), 653 mm to 742 mm (Interior)
Width (English) Interior72.5 in.
Height (English) Exterior72.6 in.
Height (English) Interior25.7 in. to 29.2 in.
Voltage230 VAC
Amperage6 A
Energy Consumption (Operating Set Point)0.37 kw
Unit SizeEach