Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™
UltraPure™ Guanidine Isothiocyanate Solution
UltraPure™ Guanidine Isothiocyanate Solution consists of 4 M guanidine isothiocyanate, 50 mM Tris-HCl (pH 7.5), 25 mM EDTA. It mayRead more
Have Questions?
Catalog number 15577018
Price (USD)/ 500 mL
509.65
Online exclusive
534.00
Save 24.35 (5%)
-
Add to cart
Price (USD)/ 500 mL
509.65
Online exclusive
534.00
Save 24.35 (5%)
Add to cart
UltraPure™ Guanidine Isothiocyanate Solution
Catalog number15577018
Price (USD)/ 500 mL
509.65
Online exclusive
534.00
Save 24.35 (5%)
-
Add to cart
UltraPure™ Guanidine Isothiocyanate Solution consists of 4 M guanidine isothiocyanate, 50 mM Tris-HCl (pH 7.5), 25 mM EDTA. It may be used as part of a protocol, based on the guanidine isothiocyanate procedure first described by Chirgwin et al. (1) and later modified by Han et al. (2), for isolating intact mRNA from tissues or cultured cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)RNA Extraction
Product TypeGuanidine Isothiocyanate
Product LineUltraPure™
PurityDNase-Free, RNase-Free
Quantity500 mL
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice