Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™
UltraPure™ Acrylamide
UltraPure™ Acrylamide is a major ingredient in polyacrylamide gels, which are used for electrophoretic separation of both nucleic acid fragmentsRead more
Have Questions?
Catalog number 15512023
Price (USD)/ 500 g
256.65
Online exclusive
268.00
Save 11.35 (4%)
-
Add to cart
Request bulk or custom format
Price (USD)/ 500 g
256.65
Online exclusive
268.00
Save 11.35 (4%)
Add to cart
UltraPure™ Acrylamide
Catalog number15512023
Price (USD)/ 500 g
256.65
Online exclusive
268.00
Save 11.35 (4%)
-
Add to cart
UltraPure™ Acrylamide is a major ingredient in polyacrylamide gels, which are used for electrophoretic separation of both nucleic acid fragments and proteins. DNA fragments generated by restriction endonuclease digestion, dideoxy sequencing, or the Maxam and Gilbert sequencing methods can be separated by polyacrylamide gel electrophoresis (PAGE). Proteins can be separated by a variety of techniques with polyacrylamide gels, including denaturing gels using SDS or urea, isoelectric focusing gels, and native gels in a wide variety of buffers. UltraPure™ Acrylamide is recrystallized three times to ensure a highly purified product.
Performance and Quality Testing: Polymerization time is evaluated in an SDS polyacrylamide gel system.