Discover the future of Gibco Media selection with our upcoming updated experience. View details >
Gibco™
KaryoMAX™ Colcemid™ Solution in PBS
Gibco™ KaryoMAX™ Colcemid™ Solution in PBS is a 10 μg/ml N-desacetyl-N-methylocolchicine (Colcemid™) solution made in Phosphate Buffered Saline (PBS). ItRead more
Have Questions?
Catalog number 15212012
Price (USD)/ 10 mL
25.63
-
Add to cart
Price (USD)/ 10 mL
25.63
Add to cart
KaryoMAX™ Colcemid™ Solution in PBS
Catalog number15212012
Price (USD)/ 10 mL
25.63
-
Add to cart
Gibco™ KaryoMAX™ Colcemid™ Solution in PBS is a 10 μg/ml N-desacetyl-N-methylocolchicine (Colcemid™) solution made in Phosphate Buffered Saline (PBS). It is used for arresting cells in metaphase so the chromosomes can be separated for cytogenetic studies and in vitro diagnostic procedures. Colcemid™ prevents spindle formation during mitosis, causing metaphase arrest. The mechanism of action is similar to that of colchicine, but with lower mammalian toxicity. The complete formulation is available.
Product Use For in vitro diagnostic use.
cGMP Manufacturing and Quality System Gibco™ KaryoMAX™ Colcemid™ Solution in PBS is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Cell TypeMammalian Cells
FormLiquid
Product TypeChromosome Analysis Media
Shelf Life18 Months
SterilitySterile-filtered
For Use With (Application)Clinical Research, Translational Research
pH6 to 8
Product LineKaryoMAX™
Quantity10 mL
Shipping ConditionRoom Temperature
Unit Size10 mL
Contents & Storage
Storage conditions: 2 – 8°C Shipping conditions: Ambient Shelf life: 18 months from date of manufacture