Discover the future of Gibco Media selection with our upcoming updated experience. View details >
Gibco™
KaryoMAX™ Colcemid™ Solution in HBSS
Gibco™ KaryoMAX™ Colcemid™ Solution in HBSS is a 10 μg/mL N-desacetyl-N-methylocolchicine (Colcemid™) solution made up in Hanks' Balanced Salt SolutionRead more
Have Questions?
Catalog number 15210040
Price (USD)/ 10 mL
26.99
-
Add to cart
Price (USD)/ 10 mL
26.99
Add to cart
KaryoMAX™ Colcemid™ Solution in HBSS
Catalog number15210040
Price (USD)/ 10 mL
26.99
-
Add to cart
Gibco™ KaryoMAX™ Colcemid™ Solution in HBSS is a 10 μg/mL N-desacetyl-N-methylocolchicine (Colcemid™) solution made up in Hanks' Balanced Salt Solution (HBSS). Colcemid™ prevents spindle formation during mitosis, arresting cells in metaphase so that the chromosomes can be separated for cytogenetic studies and in vitro diagnostic procedures. The mechanism of action is similar to that of colchicine, but with lower mammalian toxicity.
cGMP Manufacturing and Quality System Gibco™ KaryoMAX™ Colcemid™ Solution in HBSS is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Cell TypeMammalian Cells
FormLiquid
Product TypeChromosome Analysis Media
Shelf Life18 Months
SterilitySterile-filtered
For Use With (Application)Clinical Research, Translational Research
Product LineKaryoMAX™
Quantity10 mL
Shipping ConditionRoom Temperature
Unit Size10 mL
Contents & Storage
Storage conditions: 2°C to 8°C Shipping conditions: Room temperature Shelf life: 18 months from date of manufacture