Nunc™ EasYDish™ Dishes with Nunclon™ Supra Surface
Nunc™ EasYDish™ Dishes with Nunclon™ Supra Surface
Thermo Scientific™

Nunc™ EasYDish™ Dishes with Nunclon™ Supra Surface

Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Height (Metric)Culture AreaShape
150474100 mm17 mm56.7 cm2Round
15047035 mm13 mm8.8 cm2Round
15047260 mm16 mm21.5 cm2Round
150476100 mm21 mm56.7 cm2Round
150478150 mm21 mm145 cm2Round
Catalog number 150474
Price (USD)/ Case of 150
253.40
Special Offer
Online exclusive
Ends: 01-Jan-2025
362.00 
Save 108.60 (30%)
Add to cart
Diameter (Metric):
100 mm
Height (Metric):
17 mm
Culture Area:
56.7 cm2
Shape:
Round
Price (USD)/ Case of 150
253.40
Special Offer
Online exclusive
Ends: 01-Jan-2025
362.00 
Save 108.60 (30%)
Add to cart
Nunc™ EasYDish™ Dishes with Nunclon™ Supra Surface
Catalog number150474
Price (USD)/ Case of 150
253.40
Special Offer
Online exclusive
Ends: 01-Jan-2025
362.00 
Save 108.60 (30%)
-
Add to cart
Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The beveled grip ring is easy to hold and stays secure in your gloved hand. Available in four sizes: 35, 60, 100, and 150 mm. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc EasYDish Dishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

For Laboratory Use
Specifications
AutoclavableNot Autoclavable
Bottom ShapeFlat
ColorClear
Culture Area56.7 cm2
DisposableSingle-use
GriddedNot Gridded
MaterialPolystyrene
ShapeRound
SterilitySterile
TypeCell Culture Dish
No. per Case150 Dishes
No. per Pack10 Dishes
Height (Metric)17 mm
Diameter (Metric)100 mm
Unit SizeCase of 150