Catalog Number | Diameter (Metric) | Height (Metric) | Culture Area | Shape |
---|---|---|---|---|
150470 | 35 mm | 13 mm | 8.8 cm2 | Round |
150472 | 60 mm | 16 mm | 21.5 cm2 | Round |
150476 | 100 mm | 21 mm | 56.7 cm2 | Round |
150474 | 100 mm | 17 mm | 56.7 cm2 | Round |
150478 | 150 mm | 21 mm | 145 cm2 | Round |
Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.
Features of Nunc EasYDish Dishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years