Platinum™ II Green PCR Buffer (5X)
Inquire about OEM or Commercial Supply version of this product here.
Platinum™ II Green PCR Buffer (5X)
Invitrogen™

Platinum™ II Green PCR Buffer (5X)

Invitrogen 5X Platinum II Green PCR Buffer for fast and easy PCR
Have Questions?
Catalog number 14966123
Price (USD)/ 5 mL
146.00
-
Add to cart
Request bulk or custom format
Price (USD)/ 5 mL
146.00
Add to cart
Platinum™ II Green PCR Buffer (5X)
Catalog number14966123
Price (USD)/ 5 mL
146.00
-
Add to cart
Invitrogen Platinum II Green PCR Buffer is an innovative buffer optimized for use with Platinum II Taq Hot-Start DNA Polymerase. Due to its unique composition, the annealing temperature is 60°C for most primer pairs designed following the general design rules. Isostabilizing molecules in the buffer increase primer–template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize annealing temperature for each primer pair.

Platinum II Green PCR Buffer provides 1.5 mM MgCl2 in the final reaction and is supplemented with a density reagent and two tracking dyes for direct loading of PCR products on gels.

For Research Use Only.
Specifications
FormatTube
PolymerasePlatinum II Taq Hot-Start DNA Polymerase
Reaction SpeedFast
Product TypePCR Buffer
Quantity5 mL
Concentration5X
Volume4 x 1.5 mL
Unit Size5 mL
Contents & Storage
5X Platinum II Green PCR buffer, 4 x 1.25 mL

Store at -20°C in a non-frost-free freezer.