Platinum™ Hot Start PCR Master Mixes (2X)
For superior PCR performance, upgrade to Invitrogen's new Platinum II Taq Hot-start DNA Polymerase.
Inquire about OEM or Commercial Supply version of this product here.
Platinum™ Hot Start PCR Master Mixes (2X)
Invitrogen™

Platinum™ Hot Start PCR Master Mixes (2X)

Invitrogen Platinum Hot Start PCR Master Mix offers Platinum Taq DNA polymerase in an optimized PCR buffer with magnesium and dNTPs for convenient PCR setup.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. of Reactions
13000014Colorless1000 Reactions
13000012Colorless50 Reactions
13000013Colorless200 Reactions
13001012Green50 Reactions
13001013Green200 Reactions
13001014Green1000 Reactions
Catalog number 13000014
Price (USD)/ 1000 reactions
1,478.65
Online exclusive
1572.00 
Save 93.35 (6%)
-
Add to cart
Color:
Colorless
No. of Reactions:
1000 Reactions
Request bulk or custom format
Price (USD)/ 1000 reactions
1,478.65
Online exclusive
1572.00 
Save 93.35 (6%)
Add to cart
Platinum™ Hot Start PCR Master Mixes (2X)
Catalog number13000014
Price (USD)/ 1000 reactions
1,478.65
Online exclusive
1572.00 
Save 93.35 (6%)
-
Add to cart
Invitrogen Platinum Hot Start PCR Master Mix, available in colorless and green formats, offers Platinum Taq DNA polymerase in an optimized PCR buffer with magnesium and dNTPs for convenient PCR setup. It retains all the features of Platinum Taq DNA Polymerase: specificity, robustness, and reliability.

Platinum Green Hot Start PCR Master Mix additionally contains a density reagent and two tracking dyes for direct loading of PCR products on gels.

Platinum Hot Start PCR Master Mix provides a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs, thus reducing the number of pipetting steps during PCR reaction setup. It is supplied with a separate vial of Platinum GC Enhancer, formulated for specific amplification and improved yields of GC-rich targets.

Platinum Taq DNA Polymerase is a recombinant Taq DNA polymerase complexed with proprietary antibodies that inhibit enzyme activity at room temperature, reducing nonspecific target amplification at lower temperatures. The antibodies dissociate during the initial PCR denaturation step, and DNA polymerase regains its full activity. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase has a non-template-dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5' to 3' exonuclease activity.

Features

  • High specificity and increased yields with antibody-mediated hot-start PCR
  • Convenient room temperature reaction setup
  • Versatile formulation for broad range of amplicons
  • Direct gel loading with green PCR master mix

Applications

  • Hot Start PCR
  • Genotyping
  • Gene expression profiling
  • Next-generation sequencing

Notes

  • Platinum II Taq Hot-start DNA Polymerase is recommended superior PCR performance.
  • For applications that require PCR product analysis by absorbance or fluorescence excitation, colorless format is recommended.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatTube
GC-Rich PCR PerformanceMedium
PolymerasePlatinum Taq DNA Polymerase
Reaction SpeedStandard
Label or DyeUnlabeled
Product TypeHot Start PCR Master Mix
Shipping ConditionDry Ice
For Use With (Application)Hot-start PCR
Concentration2X
Fidelity (vs. Taq)1X
Hot StartBuilt-In Hot Start
No. of Reactions1000 Reactions
Overhang3'-A
Reaction FormatSuperMix or Master Mix
Size (Final Product)5 kb or less
Starting MaterialDNA
ColorColorless
Unit Size1000 reactions
Contents & Storage
• Platinum Hot Start PCR 2X Master Mix, 2 x 12.5 mL
• Platinum GC Enhancer, 10 mL
• Nuclease-free water, 2 x 12.5 mL

Store at -20°C in a non-frost-free freezer.