TrypLE™ Express Enzyme (1X), phenol red
Try TrypLE Express Enzyme (1X), phenol red for free with the Aspire member program
TrypLE™ Express Enzyme (1X), phenol red
Gibco™

TrypLE™ Express Enzyme (1X), phenol red

Green features
TrypLE Express is an animal origin-free, recombinant enzyme used for dissociating a wide range of adherent mammalian cells, including CHO,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
12605010100 mL
1260503620 x 100mL
126050931 universaL bag(s)
12605028500 mL
Catalog number 12605010
Price (USD)/ 100 mL
27.62
Special Offer
Online exclusive
Ends: 01-Jan-2025
36.82 
Save 9.20 (25%)
-
Add to cart
Quantity:
100 mL
Customize this product
Price (USD)/ 100 mL
27.62
Special Offer
Online exclusive
Ends: 01-Jan-2025
36.82 
Save 9.20 (25%)
Add to cart
TrypLE™ Express Enzyme (1X), phenol red
Catalog number12605010
Price (USD)/ 100 mL
27.62
Special Offer
Online exclusive
Ends: 01-Jan-2025
36.82 
Save 9.20 (25%)
-
Add to cart
TrypLE Express is an animal origin-free, recombinant enzyme used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A549, primary human keratinocytes, and embryonic stem cells. TrypLE Express cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Its exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts.

We offer a variety of TrypLE formulations for a range of cell culture applications.

Compared to trypsin and other dissociation reagents, Gibco TrypLE Express is:

Gentle on cells

Room-temperature stable
Easy to use
Animal origin-free (AOF)

This TrypLE Express is modified as follows:

With

• Phenol Red

• EDTA

The formulation of TrypLE solutions is available, although we keep the enzyme concentration confidential.

Gentle On Cells
TrypLE Express' exceptional purity increases specificity due to the action of a single enzyme. This reduces damage caused by cleavage from enzymes present in some trypsin preparations and in other protease extracts.

Room-Temperature Stable
TrypLE Express is room-temperature stable and ready to use. TrypLE Express remains stable for 24 months at room temperature or 2–8°C, making storage and handling easy and convenient.

Easy to Use
TrypLE Express can be directly substituted for trypsin in existing protocols. In addition, dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS.

Animal Origin-Free (AOF)
Unlike porcine trypsin, TrypLE Express is free of animal-derived components.

cGMP Manufacturing and Quality System
For supply chain continuity, we manufacture Gibco TrypLE Express at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements, are certified to the ISO 13485 standard, and are registered with the FDA as medical device manufacturers.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ChelatorsEDTA
Cell TypeMammalian Cells
ClassificationAnimal Origin-free
Concentration1 X
Culture TypeAdherent Cell Culture
FormLiquid
Product TypeCell Culture Dissociation Reagent
Shelf Life24 Months
SterilitySterile-filtered
With AdditivesPhenol Red, EDTA
Green FeaturesEnergy efficient, Sustainable packaging
OsmolalityCheck and Record
Product LineGibco™
Quantity100 mL
RemarksRoom Temperature Stable
Shipping ConditionRoom Temperature
SourceAnimal origin-free
Tested ForIn vitro Bioassay
Unit Size100 mL
Contents & Storage
Storage conditions: 15°C to 30°C, protect from light
Alternate storage conditions: 2°C to 8°C or -5°C to -20°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture