Mammalian Lumio™ Gateway™ Vectors, with Lumio™ Green In-Cell Detection Kit
Invitrogen™
Mammalian Lumio™ Gateway™ Vectors, with Lumio™ Green In-Cell Detection Kit
To fit all of your expression needs, Invitrogen offers state-of-the-art Gateway™ destination vectors for expression in E. coli, insect, yeast,Read more
Have Questions?
Catalog number 12589016
Price (USD)/ 1 kit
2,455.00
-
Add to cart
Price (USD)/ 1 kit
2,455.00
Add to cart
Mammalian Lumio™ Gateway™ Vectors, with Lumio™ Green In-Cell Detection Kit
Catalog number12589016
Price (USD)/ 1 kit
2,455.00
-
Add to cart
To fit all of your expression needs, Invitrogen offers state-of-the-art Gateway™ destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins. All Gateway™ destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is an entry clone or an Ultimate™ RF Clone. The following table lists the wide range of destination vectors available.
Additional materials required, available separately: Gateway™ entry clone, Gateway™ LR Clonase™ enzyme mix, and reaction buffer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Constitutive or Inducible SystemConstitutive
Delivery TypeTransfection
PromoterCMV
Product TypeVector with In-cell Detection Kit
Selection Agent (Eukaryotic)Blasticidin
Detection LocationIn-Cell Detection
Protein TagLumio, V5 Epitope Tag
Cloning MethodGateway
Quantity1 kit
VectorpcDNA
Product LineGateway™, Lumio™
For Use With (Application)Reporter Assays
Unit Size1 kit
Contents & Storage
All destination vectors are provided lyophilized and supercoiled.