SUMO Protease
Product Image
Invitrogen™

SUMO Protease

SUMO Protease, also known as Ulp, is a recombinant fragment of ULP1 (Ubl-specific protease 1) from Saccharomyces cerevisiae. It isRead more
Have Questions?
Catalog number 12588018
Price (USD)/ 250 units
611.65
Online exclusive
650.00 
Save 38.35 (6%)
-
Add to cart
Price (USD)/ 250 units
611.65
Online exclusive
650.00 
Save 38.35 (6%)
Add to cart
SUMO Protease
Catalog number12588018
Price (USD)/ 250 units
611.65
Online exclusive
650.00 
Save 38.35 (6%)
-
Add to cart
SUMO Protease, also known as Ulp, is a recombinant fragment of ULP1 (Ubl-specific protease 1) from Saccharomyces cerevisiae. It is highly specific for the SUMO protein fusion, recognizing the tertiary structure of SUMO rather than an amino acid sequence. The SUMO Protease features:

• Highly active cleavage
• No non-specific proteolysis (highly specific cleavage*)
• Activity from 2°C to 37°C
• A six-histidine sequence to facilitate its removal from the digested protein sample

Application: Removal of fusion tags from recombinant proteins.
Unit Definition: One unit of SUMO Protease is defined as the amount of enzyme needed to cleave 85% of 2μg of substrate protein at 30°C in one hour.
Quality Control: SUMO Protease has greater than 85% single-band purity with no non-specific protease contamination. It is functionally tested for the absence of any non-specific protease activity.
5 enzymes
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Host SpeciesS. cerevisiae
Shipping ConditionDry Ice
Quantity250 units
Unit Size250 units
Contents & Storage
SUMO Protease is provided with 10X high salt and 10X no salt dilution buffers. Store at -80°C. Guaranteed stable for 6 months when properly stored.