B-27™ Supplement (50X), minus vitamin A
B-27™ Supplement (50X), minus vitamin A
Gibco™

B-27™ Supplement (50X), minus vitamin A

A variation of the most cited neuronal cell culture supplement, B-27 Supplement Minus Vitamin A is a customized version ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
12587001100 mL
1258701010 mL
Catalog number 12587001
Price (USD)/ 100 mL
1,458.65
Online exclusive
1574.00 
Save 115.35 (7%)
-
Add to cart
Quantity:
100 mL
Price (USD)/ 100 mL
1,458.65
Online exclusive
1574.00 
Save 115.35 (7%)
Add to cart
B-27™ Supplement (50X), minus vitamin A
Catalog number12587001
Price (USD)/ 100 mL
1,458.65
Online exclusive
1574.00 
Save 115.35 (7%)
-
Add to cart
A variation of the most cited neuronal cell culture supplement, B-27 Supplement Minus Vitamin A is a customized version of the original B-27 Supplement without vitamin A (retinyl acetate). B-27 Supplement Minus Vitamin A is a serum-free supplement ideal for the cultivation of neural progenitor and stem cells, either as neurospheres in suspension or in adherent monolayer culture, without inducing differentiation. B-27 Supplement Minus Vitamin A is provided as a 50X liquid and is intended for use with Neurobasal Medium or Neurobasal-A Medium for neuronal cell culture without the need for an astrocyte feeder layer.

B-27 Supplement is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of B-27 Supplement and Neurobasal media.

For established protocols using B-27 Supplement, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeNeural Progenitor Cells
ClassificationSerum-free
Concentration50 X
Culture TypeMammalian Cell Culture
FormLiquid
Product TypeCell Culture Supplement
Serum LevelSerum-Free
Shelf Life12 Months
SterilitySterile
Green FeaturesFewer resources used, Less waste
Product LineB-27™
Quantity100 mL
Shipping ConditionDry Ice
SourceSerum-Free
Tested ForEndotoxin, Performance, Sterility
Unit Size100 mL
Contents & Storage
Store in freezer (-5 to -20°C) and protect from light.