Platinum™ PCR SuperMix High Fidelity
Inquire about OEM or Commercial Supply version of this product here.
Upgrade to Invitrogen™ Platinum™ SuperFi™ II DNA Polymerase: >300X higher fidelity than Taq, versatile and now enables universal primer annealing.
Platinum™ PCR SuperMix High Fidelity
Invitrogen™

Platinum™ PCR SuperMix High Fidelity

Platinum PCR SuperMix, High Fidelity, is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
125320245000 Reactions
12532016100 Reactions
Catalog number 12532024
Price (USD)/ 5,000 reactions
12,834.65
Online exclusive
14260.00 
Save 1,425.35 (10%)
-
Add to cart
No. of Reactions:
5000 Reactions
Request bulk or custom format
Price (USD)/ 5,000 reactions
12,834.65
Online exclusive
14260.00 
Save 1,425.35 (10%)
Add to cart
Platinum™ PCR SuperMix High Fidelity
Catalog number12532024
Price (USD)/ 5,000 reactions
12,834.65
Online exclusive
14260.00 
Save 1,425.35 (10%)
-
Add to cart
Platinum PCR SuperMix, High Fidelity, is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification. Designed for high-specificity, high-fidelity DNA amplification, Platinum PCR SuperMix reduces reaction set-up time and only requires the addition of template and primer.

High fidelity is provided by a mixture of Taq DNA polymerase and the proofreading Pyrococcus species GB-D polymerase. High specificity is achieved by anti-Taq antibodies, allowing for automatic hot-start PCR.

Benefits of Platinum PCR SuperMix, High Fidelity

  • Six-times higher fidelity than Taq DNA polymerase
  • Amplifies fragments up to 15 kb
  • Minimizes PCR optimization
  • Room-temperature reaction assembly

Applications

  • High-fidelity PCR
  • Cloning
  • Mutagenesis

Notes

  • Platinum PCR SuperMix, High Fidelity, produces products that are mixed blunt and 3-A' ends; however, the majority of ends will have a 3-A' overhangs.
  • Hot-start capability increases specificity and yield and allows for room-temperature reaction assembly.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
GC-Rich PCR PerformanceLow
PolymerasePlatinum Taq DNA Polymerase High Fidelity
Reaction SpeedStandard
Shipping ConditionWet or Dry Ice
For Use With (Application)Hot-start PCR, High-fidelity PCR
Concentration1.1X
Fidelity (vs. Taq)6X
Hot StartBuilt-In Hot Start
No. of Reactions5000 Reactions
OverhangMixed
Reaction FormatSuperMix or Master Mix
Size (Final Product)15 kb or less
Unit Size5,000 reactions
Contents & Storage
Platinum PCR SuperMix High Fidelity (4 x 56.25 mL)

Store at -20°C in a non-frost-free freezer.