Phadia™ 1000 Immunoassay Analyzers
Phadia™ 1000 Immunoassay Analyzers
Thermo Scientific™

Phadia™ 1000 Immunoassay Analyzers

Provide an optimal allergy testing solution using advanced, state-of-the-art technology with the Phadia™ 1000 Laboratory System.
Have Questions?
Catalog number 12380001
Price (USD)/ Each
-
Request A Quote
Throughput:
1500 results per 8 hour shift
Width (English):
71 in.
Depth (English):
40 in.
Height (English):
65 in.
Provide optimal allergy testing solutions using advanced, state-of-the-art technology. The Phadia™ 1000 Laboratory System is the optimal solution for the large, cost-conscious laboratory to meet the growing need for allergy testing. Phadia 1000 is designed for high-throughput laboratories, delivering 2,000 to 10,000 results per week, with proven performance in laboratories worldwide.
Delivering the first result in 100 minutes and then one result every 15 seconds, Phadia 1000 is designed for the high-volume laboratory delivering approximately 2,000 to 10,000 tests per week.
Phadia 1000 can be connected to a Laboratory Automation System for fast and reliable transportation of samples. All instruments of the Phadia Laboratory Systems family can be clustered, allowing laboratories to share resources, and thereby ensure that samples are processed as efficiently as possible.
High Productivity and Automations
  • Throughput: >1200 results in one assay
  • Fully Automated processing of ImmunoCAP assays
  • Continuous Random Access
  • Onboard reagent storage, up to 10,000 tests and all reagents
  • Positive identification and full traceability of all samples and reagents
  • Accurate, standardized and reproducible test results
  • True quantitative measurements

Cost Effectiveness
  • Large panel of standardized, high-quality allergens, allergen components and antigens available
  • 40μl serum or plasma needed per test, plus dead volume depending on the sample tube
  • 28-day storage of calibration curves

Remote diagnostics
The ImmunoCAP Specific IgE blood test assay portfolio covers a wide range of tests including: grass, weed and tree pollens, microorganisms, mites, cats, dogs and other furred animals, insects, venoms and food allergens.
Throughput and Productivity
Phadia systems are all highly automated, requiring minimal hands-on time. Choosing the system with the capacity matching your needs for throughput ensures optimal productivity.
Specialized Software
Phadia Information Data Manager (IDM) Software
  • Allows 28 days storage of standard curves
  • Allows export of results in several formats for connectivity with Laboratory Information Systems (LIS)
  • Offers flexible report options
  • Offers multilingual support

Remote Diagnostics
Phadia LabCommunity provides customers with enhanced support and troubleshooting assistance that reduces system downtime. Features include:
  • Remote collection of data needed for service and support activities
  • Proactive system health status check
  • Automatic reporting of Quality Club results to Phadia
Specifications
TypePhadia 1000 Immunoassay Analyzer
Throughput1500 results per 8 hour shift
Ambient Temperature18°C to 32°C
Depth (English)40 in.
Height (English)65 in.
Width (English)71 in.
Voltage230 V
Mainframe Connection ProtocolASTM, MasterCAP™, Phamas™
Max. Networked Workstations5 systems of each type
Operating SystemWindows™2000/ Windows XP
Weight (English)1700 lb.
Unit SizeEach