Platinum™ SuperFi II DNA Polymerase
Inquire about OEM or Commercial Supply version of this product here.
Platinum™ SuperFi II DNA Polymerase
Invitrogen™

Platinum™ SuperFi II DNA Polymerase

Invitrogen Platinum SuperFi II DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with an innovative buffer, enabling universal primer annealing for the highest success in PCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
12361050500 Reactions
12361010100 Reactions
123612505 x 500 Reactions
Catalog number 12361050
Price (USD)/ 500 reactions
1,068.65
Online exclusive
1188.00 
Save 119.35 (10%)
-
Add to cart
No. of Reactions:
500 Reactions
Request bulk or custom format
Price (USD)/ 500 reactions
1,068.65
Online exclusive
1188.00 
Save 119.35 (10%)
Add to cart
Platinum™ SuperFi II DNA Polymerase
Catalog number12361050
Price (USD)/ 500 reactions
1,068.65
Online exclusive
1188.00 
Save 119.35 (10%)
-
Add to cart
Invitrogen Platinum SuperFi II DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with an innovative buffer, enabling universal primer annealing for the highest success in PCR. Due to the unique composition of the SuperFi II PCR buffer, the annealing temperature is 60°C for most primer pairs designed following the general design rules.

Features of Platinum SuperFi II DNA Polymerase include:

  • Exceptional >300X Taq fidelity
  • Universal primer annealing at 60°C
  • Superior specificity, sensitivity, and yields
  • Robust amplification of difficult-to-amplify targets

Platinum SuperFi II DNA Polymerase is an engineered enzyme with high processivity and increased resistance to PCR inhibitors. It also enables fast-cycling protocols and amplification of long targets (up to 20 kb). Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation. This technology also enables reaction setup at room temperature and provides increased sensitivity and yield.

Isostabilizing molecules in the buffer increase primer-template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize the annealing temperature for each primer pair. With Platinum SuperFi II DNA Polymerase, different PCR assays can be cycled together using the same protocol with a universal primer annealing temperature and an extension step selected for the longest fragment to be amplified.

Applications

  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates
  • Template generation for sequencing
  • High-throughput PCR
  • Amplification of samples with suboptimal purity
  • Long PCR
Specifications
GC-Rich PCR PerformanceHigh
PolymerasePlatinum SuperFi II DNA Polymerase
Reaction SpeedFast
Shipping ConditionDry Ice
For Use With (Application)Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq)>300X
Hot StartBuilt-In Hot Start
No. of Reactions500 Reactions
OverhangBlunt
Reaction FormatStandalone
Size (Final Product)20 kb or less
Unit Size500 reactions
Contents & Storage
• Platinum SuperFi II DNA Polymerase (500 μL)
• 5X SuperFi II Buffer (6 x 1.25 mL)

Store at –20°C.