The SampleRack (for DynaMag™-5) is the same tube rack that is supplied with the DynaMag™-5 magnet. It is suitable asRead more
Have Questions?
Catalog number 12304D
Price (USD)/ 8 samples
312.65
Online exclusive
395.00
Save 82.35 (21%)
-
Add to cart
Price (USD)/ 8 samples
312.65
Online exclusive
395.00
Save 82.35 (21%)
Add to cart
SampleRack (for DynaMag™-5 magnet)
Catalog number12304D
Price (USD)/ 8 samples
312.65
Online exclusive
395.00
Save 82.35 (21%)
-
Add to cart
The SampleRack (for DynaMag™-5) is the same tube rack that is supplied with the DynaMag™-5 magnet. It is suitable as a replacement or supplement to the SampleRack provided with the DynaMag™-5 magnet. Additional SampleRacks may be useful when there are multiple users of the DynaMag™-5 magnet or when processing more than 8 samples at a time.
The SampleRack (for DynaMag™-5) is intended for use with the magnet base of the DynaMag™-5 magnet. The SampleRack holds up to 8 cell samples at a time, with numbered positions (1 to 8) for easy control of your samples and a design that ensures good sample visibility when placed on the magnet base (not supplied).
The flexible clips that hold the tubes in place allow the SampleRack to accommodate tubes from 11–16 mm in diameter, such as most standard flow tubes and blood collection tubes (Vacutainer™ tubes), with an optimal working volume of 1–5 mL.
Note: The SampleRack (for DynaMag™-5) should not be used with standard 15 mL tubes; for this purpose use DynaMag™-15 magnet.
For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.