Specifications
Certifications/ComplianceThese incubators are manufactured in accordance with CE guidelines, and meet the requirements of IEC 6110.
DescriptionCell Factory Incubator
MaterialStainless Steel
Product TypeIncubator
Temperature Range (Metric)5° to 50°C
Voltage208/240V
Frequency50/60Hz
Electrical Requirements208/240V 50/60Hz
Unit SizeEach