Ion Source Replacement, spare GD source
Ion Source Replacement, spare GD source
Thermo Scientific™
Ion Source Replacement, spare GD source
Gain superior sensitivity, short analysis times and low interference levels, with this fast flow, high power glow discharge ion source.
Have Questions?
Catalog number 1176403
Price (USD)/ Each
-
Request A Quote
Description:
Spare GD source
Ensure ongoing peak performance from your Thermo Scientific™ ELEMENT GD™ GD-MS analyzer; use only the highest quality, dedicated replacement parts, such as this Thermo Scientific™ Ion Source Replacement.
With this fast flow, high power glow discharge ion source, you gain superior sensitivity, short analysis times and low interference levels.
Specifications
TypeIon Source Replacement
PowerDC Voltage: Adjustable, Max. 1,400 V, Working Range Typical 400-1,200 V Current: Adjustable, Max. 200 mA, Typical 30-50 mA Pulsed DC Option with 10 to 150 μs Pulse Duration Peltier Cooling of The Sample
DescriptionSpare GD source
For Use With (Equipment)ELEMENT GD GD-MS Instrument
Unit SizeEach