Model 1150 Dual Reactor
Model 1150 Dual Reactor
Thermo Scientific™
Model 1150 Dual Reactor
Reliably remove carbon monoxide and hydrocarbons from filtered ambient air using dual reactor technology.
Have Questions?
Catalog number 1150-EPD
Price (USD)/ Each
-
Request A Quote
Description:
Model 1150 Dual Reactor
Reliably remove carbon monoxide and hydrocarbons from filtered ambient air using two reactors in series, providing efficient catalytic conversion of CO and HC to CO2 and H2O at ∽350°C with the Thermo Scientific™ Model 1150 Dual Reactor.
When utilized for carbon monoxide removal in conjunction with the Thermo Scientific Model 48i Carbon Monoxide Analyzer, the air from a compressor that passes through the Model 1150 Dual Reactor can be used for dilution or zero air. When utilized for hydrocarbon, methane and non-methanes removal in conjunction with the Thermo Scientific Model 51i Analyzer or 55i Analyzer, the air can be utilized as make-up air or zero air. A 0-100 PSIG regulator and pressure gauge are also provided to reduce the output pressure to a desired flow and pressure rate.
Specifications
Detectable AnalytesCarbon Monoxide, Hydrocarbons
Flow Rate0 to 20 L/min.
Certifications/ComplianceUKCA
DescriptionModel 1150 Dual Reactor
Electrical Requirements115 VAC/60 Hz; 220 VAC/50 Hz
Frequency50/60 Hz
Outputs< 0.1ppm CO or HC
PressureUp to 100 PSIG
Unit SizeEach