Dynabeads™ FlowComp™ Human CD14 Kit
Dynabeads™ FlowComp™ Human CD14 Kit
Invitrogen™

Dynabeads™ FlowComp™ Human CD14 Kit

Dynabeads™ FlowComp™ Human CD14 contain magnetic beads that provide a robust and versatile tool to positively isolate pure CD14+ monocytesRead more
Have Questions?
Catalog number 11367D
Price (USD)/ 2 x 3 mL
1,406.00
-
Add to cart
Price (USD)/ 2 x 3 mL
1,406.00
Add to cart
Dynabeads™ FlowComp™ Human CD14 Kit
Catalog number11367D
Price (USD)/ 2 x 3 mL
1,406.00
-
Add to cart
Dynabeads™ FlowComp™ Human CD14 contain magnetic beads that provide a robust and versatile tool to positively isolate pure CD14+ monocytes from whole blood, bone marrow, buffy coat, mononuclear cells (MNC), or tissue digests. The cells can be released from the beads using the FlowComp™ Release Buffer.
• Rapid isolation of CD14+ monocytes from any sample – no columns required
• Pure and bead-free CD14+ monocytes for any downstream application

Excellent Yield, Purity, and Viability of Recovered Cells
Dynabeads™ FlowComp™ Human CD14 contains uniform, superparamagnetic beads (2.8 μm diameter) that enable easy isolation of human CD14+ monocytes directly from any sample via a three-step process. First, the FlowComp™ Human CD14 Antibody is mixed with the sample in a tube where it binds to the CD14+ monocytes during a short incubation. Then, the CD14+ monocytes that have bound the specific antibodies are captured by the FlowComp™ Dynabeads™ and separated from the unbound cells using a magnet. Finally, the CD14+ monocytes are released from the beads by adding the FlowComp™ Release buffer. This fast and gentle isolation method does not require the use of columns, and helps ensure high purity and viability of the isolated CD14+ monocytes.

Select what is right for you from our range of Human Monocyte Cell Isolation Products.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMonocytes
ClonalityMonoclonal
Product TypeSuperparamagnetic Beads
Final Product TypeCells
FormatBeads in solution
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Isolation TechnologyPositive isolation - cells are left bead-free after isolation
Ligand TypeAntibody
No. of Cells2 x 109 PBMC, 80 mL whole blood, or 160 mL buffy coat
No. of Tests40 Isolations
Product LineFlowComp™
Purity or Quality GradeResearch Grade, 99%
Quantity2 x 3 mL
Sample TypePBMC, Buffy Coat, Blood
Shipping ConditionRoom Temperature
Target SpeciesHuman
Diameter (Metric)2.8 μm
Unit Size2 x 3 mL
Contents & Storage
The Kit Contains:
• 2 x 3 mL FlowComp™ Dynabeads™
• 1 mL FlowComp™ Human CD14 Antibody
• 2 x 20 mL FlowComp™ Release Buffer.

Store at: 2°C to 8°C.