Dynabeads™ FlowComp™ Human CD3 Kit
Dynabeads™ FlowComp™ Human CD3 Kit
Invitrogen™

Dynabeads™ FlowComp™ Human CD3 Kit

Dynabeads™ FlowComp™ Human CD3 contain magnetic beads that provide a robust and versatile tool to positively isolate pure CD3+ T-cellsRead more
Have Questions?
Catalog number 11365D
Price (USD)/ 1 kit
1,110.65
Online exclusive
1406.00 
Save 295.35 (21%)
-
Add to cart
Price (USD)/ 1 kit
1,110.65
Online exclusive
1406.00 
Save 295.35 (21%)
Add to cart
Dynabeads™ FlowComp™ Human CD3 Kit
Catalog number11365D
Price (USD)/ 1 kit
1,110.65
Online exclusive
1406.00 
Save 295.35 (21%)
-
Add to cart
Dynabeads™ FlowComp™ Human CD3 contain magnetic beads that provide a robust and versatile tool to positively isolate pure CD3+ T-cells from whole blood, bone marrow, buffy coat, mononuclear cells (MNC), or tissue digests. The cells can be released from the beads using the FlowComp™ Release Buffer.
• Rapid isolation of CD3+ T-cells from any sample – no columns required
• Pure and bead-free CD3+ T-cells for any downstream application

Excellent Yield, Purity, and Viability of Recovered Cells
Dynabeads™ FlowComp™ Human CD3 contains uniform, superparamagnetic beads (2.8 μm diameter) that enable easy isolation of human CD3+ cells directly from any sample via a three-step process. First, the FlowComp™ Human CD3 Antibody is mixed with the sample in a tube where it binds to the target cells during a short incubation. Then, the CD3+ T-cells that have bound the specific antibodies are captured by the FlowComp™ Dynabeads™ and separated from unbound cells using a magnet. Finally, the target cells are released from the beads by adding the FlowComp™ Release buffer. This fast and gentle isolation method does not require the use of columns, and helps ensure high purity, recovery and viability of the isolated CD3+ cells. The cells can be used in any downstream application.

Select what is right for you from our range of Human T Cell Isolation Products.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeT Cells (CD3)
Product TypeSuperparamagnetic Beads
Final Product TypeCells
FormatBead
Isolation TechnologyPositive Isolation
No. of Cells80 mL whole blood, or 2 x 109 PBMC
Product LineDYNAL™, Dynabeads™, FlowComp™
Purity or Quality GradeResearch Grade, 99%
Quantity1 kit
Sample TypeMNC, Blood
Shipping ConditionRoom Temperature
Target SpeciesHuman
Diameter (Metric)2.8 μm
Unit Size1 kit
Contents & Storage
The Kit Contains:
• 1 mL Antibody Mix
• 3 mL FlowComp™ Dynabeads™
• 40 mL FlowComp™ Release Buffer.

Store at: 2°C to 8°C.