Dynabeads™ Biotin Binder
Dynabeads™ Biotin Binder
Invitrogen™

Dynabeads™ Biotin Binder

Dynabeads™ Biotin Binder are 2.8 μm superparamagnetic beads that bind biotinylated antibodies (or other biotinylated ligands), allowing you to isolateRead more
Have Questions?
Catalog number 11047
Price (USD)/ 5 mL
650.65
Online exclusive
824.00 
Save 173.35 (21%)
-
Add to cart
Price (USD)/ 5 mL
650.65
Online exclusive
824.00 
Save 173.35 (21%)
Add to cart
Dynabeads™ Biotin Binder
Catalog number11047
Price (USD)/ 5 mL
650.65
Online exclusive
824.00 
Save 173.35 (21%)
-
Add to cart
Dynabeads™ Biotin Binder are 2.8 μm superparamagnetic beads that bind biotinylated antibodies (or other biotinylated ligands), allowing you to isolate cells from any species (e.g., human, mouse, rat, non-human primates, and cell lines).

• Works with any cell type, from any sample and from any species
• Fast and efficient depletion or positive isolation of any target cells for molecular downstream assays using a biotinylated primary antibody
• Obtain pure and untouched cells using a mixure of biotinylated antibodies to deplete unwanted cells

Adaptable for various isolation strategies
The biotinylated primary antibody may be added to the cell sample (indirect technique) or pre-coated onto the Dynabeads™ Biotin Binder (direct technique). Regardless of which of these techniques is used, when the cell sample and the Dynabeads™ Biotin Binder are mixed, the beads bind to the target cells. Placing the sample on a magnet separates the bead-bound cells from the rest of the sample. One or more biotinylated antibodies may be used to either deplete the sample of unwanted cells (negative isolation) or to capture specific cells for downstream experiments (positive isolation). For isolation of bead-free cells (e.g., for flow cytometry), we recommend using your biotinylated antibody in combination with the CELLection™ Biotin Binder Kit.

Useful for many sample types
Using Dynabeads™ Biotin Binder, you can isolate cells basically from any species, and from many starting samples such as whole blood, cord blood, peripheral blood mononuclear cells (PBMC), buffy coat, mouse spleen/lymph nodes, and tissue digests.

Learn more about Dynabeads™ products
• Find Dynabeads™ products for a whole range of applications.
• Find magnets for Dynabeads™ separations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeAll cells from all species
Product TypeBiotin Binder
Isolation TechnologyDepletion, negative isolation or positive isolation
No. of CellsProcesses ∼2 x 109 cells total
Output Viability>95%
Product LineDYNAL™, Dynabeads™
Purity or Quality GradeResearch Grade
Quantity5 mL
ReactivityAll species
Sample TypePBMC, Tissue Digests, Blood
Shipping ConditionRoom Temperature
Starting Material Cell No.1 x 107 PBMCs per isolation
Target SpeciesAll species
Diameter (Metric)2.8 μm
Unit Size5 mL
Contents & Storage
This Product Contains: 5 mL Dynabeads™ coated with recombinant streptavidin.

Store at 2°C to 8°C.