Specifications
pH6 to 8
Recommended Storage• 1 bottle of UltraPure DNase/RNase-Free Distilled Water (500 mL/bottle)
Store at room temperature.
GradeMolecular Biology
PackagingBottle
Treatment(s)Not DEPC-Treated
Quantity500 mL
Purification MethodMembrane-Filtered
Shipping ConditionRoom Temperature
Product LineUltraPure
Green FeaturesSustainable packaging
Unit Size500 mL