Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™
High DNA Mass Ladder
Invitrogen High DNA Mass Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 1,000Read more
Have Questions?
Catalog number 10496016
Price (USD)/ 26 µg
140.65
Special Offer
Online exclusive
Ends: 16-Dec-2024
152.00
Save 11.35 (7%)
-
Add to cart
Request bulk or custom format
Price (USD)/ 26 µg
140.65
Special Offer
Online exclusive
Ends: 16-Dec-2024
152.00
Save 11.35 (7%)
Add to cart
High DNA Mass Ladder
Catalog number10496016
Price (USD)/ 26 µg
140.65
Special Offer
Online exclusive
Ends: 16-Dec-2024
152.00
Save 11.35 (7%)
-
Add to cart
Invitrogen High DNA Mass Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 1,000 bp to 10,000 bp. High DNA Mass Ladder consists of six individual chromatography-purified DNA fragments.
High DNA Mass Ladder is ideal for separation on 0.8–1% agarose gels.
Highlights of High DNA Mass Ladder: • Sharp, clear bands—chromatography purified fragments for consistent and reliable results • Convenient—provided with 10X BlueJuice Gel Loading Buffer for tracking of sample DNA migration • Precise—an exact amount of DNA in each band
Product use The double-stranded DNA ladder can be visualized on 0.8–1% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with an equimolar amount of DNA in each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.13 μg/μL
Gel TypeAgarose
Product TypeHigh DNA Mass Ladder
Ready to LoadNo
Sample Loading Volume1 mL
Size Range1000 to 10000 bp
Volume (Metric)200 μL
Gel CompatibilityAgarose gel
Green FeaturesSustainable packaging
No. of Reactions50 Applications
Quantity26 μg
Shipping ConditionApproved for shipment at Room Temperature or on Dry Ice
TechnologyIndividual chromatography-purified DNA fragments
Unit Size26 µg
Contents & Storage
• 200 µL High DNA Mass Ladder • 1 mL10X BlueJuice Gel Loading Buffer