E-Gel™ Sizing DNA Ladder
E-Gel™ Sizing DNA Ladder
Invitrogen™

E-Gel™ Sizing DNA Ladder

Green features
Invitrogen E-Gel Sizing DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 50Read more
Have Questions?
Catalog number 10488100
Price (USD)/ 100 applications
239.65
Online exclusive
248.00 
Save 8.35 (3%)
-
Add to cart
Price (USD)/ 100 applications
239.65
Online exclusive
248.00 
Save 8.35 (3%)
Add to cart
E-Gel™ Sizing DNA Ladder
Catalog number10488100
Price (USD)/ 100 applications
239.65
Online exclusive
248.00 
Save 8.35 (3%)
-
Add to cart
Invitrogen E-Gel Sizing DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 50 to 1,500 bp. E-Gel Sizing DNA Ladder consists of 19 individual chromatography-purified DNA fragments with reference bands at 1000, 500, and 200 bp for easy orientation.

E-Gel Sizing DNA Ladder is specifically formulated for optimal performance on pre-cast E-Gel agarose gels.

Highlights of E-Gel Sizing DNA Ladder:
Performance—optimized for use on E-Gel SizeSelect ll agarose gels
Sharp, clear bands—chromatography-purified fragments for consistent and reliable results, stable at room temperature for up to six months
Ready to use—premixed with loading buffer
Convenient—provided with 1X E-Gel Sample Loading Buffer
Precise—an exact amount of DNA in each band

Product use
The double-stranded DNA ladder can be visualized on 2% E-Gel SizeSelect II and E-Gel EX agarose gels. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.002 μg/μL
Gel TypeAgarose
Product TypeDNA Ladder
Ready to LoadYes
Sample Loading Volume1.25 mL
Shelf Life6 Months
Size Range50 to 1500 bp
Volume (Metric)2 x 1.25 mL
Gel CompatibilitySingle Comb E-Gels, SizeSelect™ II E-Gels
Green FeaturesSustainable packaging
Quantity100 applications
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
TechnologyIndividual chromatography-purified DNA fragments
Unit Size100 applications
Contents & Storage
• 2 x 1.25 mL E-Gel Sizing DNA Ladder
• 1.25 mL 1X E-Gel Sample Loading Buffer

Store at room temperature or at 4°C for up to 6 months.
For longer periods store at -20°C.