DRI™ Ecstasy Plus Drugs of Abuse Assay
Product Image
Thermo Scientific™

DRI™ Ecstasy Plus Drugs of Abuse Assay

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Ecstasy Plus Drugs of Abuse Assay.
Have Questions?
Catalog number 10024631
Price (USD)/ Each
-
Request A Quote
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Ecstasy Plus Drugs of Abuse Assay. Supports new SAMHSA guidelines (effective October 1, 2017).
  • Supports new SAMHSA guidelines (effective October 1, 2017)
  • Detects MDMA at 100%
  • Detects MDA with cross-reactivity of ≥80%
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionSupports new SAMHSA guidelines (effective October 1, 2017).
Detectable AnalytesMDA, MDEA, MDMA
DoA CalibratorsDRI MDA Control Kit
TypeReagent
Quantity1 x 500 mL
Shelf LifeUntil Label Expiry Date
StabilityUntil label expiry date
Unit SizeEach
Contents & Storage
2°C to 8°C