CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators
CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators
Thermo Scientific™
CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators
Achieve optimal low-end sensitivity and a broad dynamic range with the Thermo Scientific™ CEDIA™ Cyclosporine Plus Immunoassay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
100147CEDIA Cyclosporine PLUS Combo Kit
100012CEDIA Cyclosporine (CsA) PLUS High Calibrator Set
Catalog number 100147
Price (USD)/ Each
-
Request A Quote
Description:
CEDIA Cyclosporine PLUS Combo Kit
Achieve precise and accurate quantitative results crucial for monitoring drug levels of cyclosporine in kidney, liver and heart transplant patients with our easy-to-use Thermo Scientific™ CEDIA™ Cyclosporine Plus Immunoassay. Used with the Thermo Scientific CEDIA Plus High Range Calibrators, this immunoassay achieves a broad dynamic range of 25-2000ng/mL for both trough and peak samples. Applications are available on our easy-to-use Indiko™ and Indiko Plus clinical and specialty chemistry systems as well as a wide variety of other laboratory instrumentation.
Key Features
  • Optimal low-end sensitivity with a broad dynamic range
  • Excellent correlation with HPLC-MS, the gold standard method
  • Precise throughout the standard curve assay range
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionCEDIA Cyclosporine PLUS Combo Kit
Detectable AnalytesCyclosporine (CsA)
Sample TypePlasma, Serum, Plasma, Serum
TypeReagent
Quantity41 mL, 19 mL
Unit SizeEach
Contents & Storage
2°C to 8°C