Attune™ NxT No-Wash No-Lyse Filter Kit
Attune™ NxT No-Wash No-Lyse Filter Kit
Invitrogen™

Attune™ NxT No-Wash No-Lyse Filter Kit

The Attune™ NxT No-Wash No-Lyse Filter Kit enables configuration of the Attune™ NxT Acoustic Focusing Cytometer with an additional sideRead more
Have Questions?
Catalog number 100022776
Price (USD)/ 1 kit
1,139.65
Online exclusive
1308.00 
Save 168.35 (13%)
-
Add to cart
Price (USD)/ 1 kit
1,139.65
Online exclusive
1308.00 
Save 168.35 (13%)
Add to cart
Attune™ NxT No-Wash No-Lyse Filter Kit
Catalog number100022776
Price (USD)/ 1 kit
1,139.65
Online exclusive
1308.00 
Save 168.35 (13%)
-
Add to cart
The Attune™ NxT No-Wash No-Lyse Filter Kit enables configuration of the Attune™ NxT Acoustic Focusing Cytometer with an additional side scatter channel off of the violet laser, allowing no-lyse no-wash immunophenotyping of whole blood. The Attune™ NxT cytometer is quickly and easily configured to collect violet side scatter by changing dichroic and emission blades.

Enables no-wash/no-lyse application
Immunophenotyping of whole blood using traditional flow cytometry is typically performed with lysis of the red blood cells (RBCs) followed by a wash step. These steps can result in significant cell loss and damage.

While a no-lyse, no-wash method for immunophenotyping of whole blood has been previously described, the method is of limited utility in traditional flow cytometry, which use hydrodynamic forces to orient cells in the flow stream. Moreover, significant sample dilution is required in no-lyse, no-wash methods to achieve low coincidence with red blood cells, necessitating extremely long acquisition times.

With the Attune™ NxT cytometer you can acquire data quickly, even from dilute samples, using either a fluorescent reagent as a trigger to eliminate red blood cells or a differential side scatter using both blue and violet scatter. This kit enables the side scatter approach.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAttune™
Quantity1 kit
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeNo-Wash No-Lyse Filter Kit
Unit Size1 kit
Contents & Storage
Contains:
• 1 Dichroic 415 LP filter
• 1 Emission 405/10 BP filter

Store (at room temperature.)