Dionex™ ASE™ Prep Sorbent Cartridges
Dionex™ ASE™ Prep Sorbent Cartridges
Actual product may vary
Thermo Scientific™
Dionex™ ASE™ Prep Sorbent Cartridges
Obtain lower GC, GC/MS, or LC/MS backgrounds with fewer interfering substances by using these cartridges for post-extraction cleanup of organic contaminants from solid and semi-solid matrices.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
088428Dionex ASE Prep Florisil Cartridges
088430Dionex ASE Alumina Neutral Cartridges
088434Dionex ASE Alumina Base Cartridges
088433Dionex ASE Alumina Acid Cartridges
Catalog number 088428
Price (USD)/ Pack of 30
225.00
Add to cart
Type:
Dionex ASE Prep Florisil Cartridges
Price (USD)/ Pack of 30
225.00
Add to cart
Dionex™ ASE™ Prep Sorbent Cartridges
Catalog number088428
Price (USD)/ Pack of 30
225.00
-
Add to cart
Chromatograms show lower backgrounds and fewer interfering substances after cleanup using Thermo Scientific™ Dionex™ ASE™ Prep Sorbent Cartridges. Use these cartridges for post-extraction cleanup of organic contaminants from solid and semi-solid matrices typically obtained from environmental, biofuel, and food and beverage industries. As part of a workflow, Dionex ASE Prep Sorbent Cartridges can be used after extraction on the Dionex ASE Accelerated Solvent Extractor and before to GC, GC/MS and LC/MS analysis.
Easy to use
Both the analytes of interest and the matrix interferences are retained and concentrated on the cartridge. The analytes are eluted with a weak solvent while polar matrix interferences remain on the cartridge.
Choose from four chemistries
Dionex ASE Prep Florisil Cartridges
  • A selective synthetic form of magnesium silicate and a normal-phase sorbent
  • Removes polar interferences from extracts prior to GC or GC-MS analysis
Dionex ASE Alumina Neutral Cartridges
  • Concentrates aldehydes, ketones, quinones, esters, lactones, and glycosides from extracts prior to GC or GC-MS analysis
Dionex ASE Alumina Base Cartridges
  • Concentrates polar analytes including pesticides, herbicides, and organic pollutants from extracts prior to GC or GC-MS analysis
Dionex ASE Alumina Acid Cartridges
  • Concentrates polar analytes such as vitamins from extracts prior to GC or GC-MS analysis.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description<ul><li> Particle Size: 60-100 Mesh</li><li>Surface Area: 158 m2/g</li><li>Shelf Life : 5 years</li></ul>
Quantity30 Pk.
Volume (Metric)6.0 mL
TypeDionex ASE Prep Florisil Cartridges
Unit SizePack of 30