B·R·A·H·M·S™ PAPP-A KRYPTOR™
B·R·A·H·M·S™ PAPP-A KRYPTOR™
Actual product may vary
Thermo Scientific™
B·R·A·H·M·S™ PAPP-A KRYPTOR™
B·R·A·H·M·S provides a complete portfolio of biomarkers for the most reliable results in prenatal screening. Reach outstanding precision and lot-to-lot long term stability for trisomy and pre-eclampsia screening in first trimester of pregnancy with B·R·A·H·M·S PAPP-A KRYPTOR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Application)
086691NCalibrator for B·R·A·H·M·S PAPP-A KRYPTOR (1 level)Readjust the standard curve memorized in B·R·A·H·M·S KRYPTOR compact PLUS/KRYPTOR GOLD
866075NImmunofluorescent assay for quantitative determination of Pregnancy Associated Plasma Protein A (PAPP-A)Risk assessement of fetal trisomy 21, 18 and 13 in first trimester of pregnancy, risk assessment of pre-eclampsia in first trimester of pregnancy
Catalog number 086691N
Price (USD)/ Box with 6 vials
-
Request A Quote
Description:
Calibrator for B·R·A·H·M·S PAPP-A KRYPTOR (1 level)
For Use With (Application):
Readjust the standard curve memorized in B·R·A·H·M·S KRYPTOR compact PLUS/KRYPTOR GOLD
B·R·A·H·M·S provides a complete portfolio of biomarkers for the screening of trisomy in first and second trimester of pregnancy and the management of pre-eclampsia. B·R·A·H·M·S PAPP-A KRYPTOR can be used as an aid for early identification of the pregnancies at risk of trisomy 21, 18 and 13 in first trimester and the early identification in the first trimester of the women at risk of developing pre-eclampsia.
US Only: Product is not 510k cleared in US.
Get confidence in your results with the high precision and the low level of interference of B·R·A·H·M·S PAPP-A KRYPTOR.
  • Save cost and time with a multiparametric control containing 3 parameters: PAPP-A, Free βhCG and AFP
  • Incubation time of only 19 minutes for a short turnaround time
  • Allows measurement in serum
  • Large capacity with 75 tests
  • Kit stable up to 29 days after opening onboard the instrument
  • Easy workflow with only 1 calibrator and one calibration every 2 weeks
  • Ready for use reagents
  • Fits your throughput need with B·R·A·H·M·S KRYPTOR compact PLUS or B·R·A·H·M·S KRYPTOR GOLD
  • Allows risk calculation in first trimester with B·R·A·H·M·S Fast screen pre I plus
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
Specifications
DescriptionCalibrator for B·R·A·H·M·S PAPP-A KRYPTOR (1 level)
Detectable AnalytesPAPP-A
For Use With (Application)Readjust the standard curve memorized in B·R·A·H·M·S KRYPTOR compact PLUS/KRYPTOR GOLD
IncludesCalibrator kit, 6 vials
Analysis Time19 minutes
FormReady-to-Use (RTU)
Unit SizeBox with 6 vials
Contents & Storage
Calibrator kit, 6 vials, 2°C to 8°C