Dionex™ ASE™ Prep MAP
Dionex™ ASE™ Prep MAP
Thermo Scientific™
Dionex™ ASE™ Prep MAP
Used with Thermo Scientific™ Dionex™ ASE systems, this unique formulation allows quick, effective water removal independent of the ionic strength of the matrix.
Have Questions?
Catalog number 083475
Price (USD)/ Each 1
144.00
Add to cart
Volume (Metric):
200 g
For Use With (Equipment):
Dionex ASE Systems
Price (USD)/ Each 1
144.00
Add to cart
Dionex™ ASE™ Prep MAP
Catalog number083475
Price (USD)/ Each 1
144.00
-
Add to cart
Designed for use with Thermo Scientific™ Dionex™ ASE™ Accelerated Solvent Extractor systems, this unique formulation of Dionex ASE Prep MAP Dispersant and Sorbent allows quick, effective water removal independent of the ionic strength of the sample matrix. The proprietary MAP polymer is a copolymer of an anionic and cationic monomer. It is a free-flowing white material and can be easily mixed with ASE Prep DE (diatomaceous earth) in a 1:1 ratio for optimum moisture removal under accelerated solvent extraction conditions.
Dionex ASE Accelerated Solvent Extractor Systems
Dionex ASE systems automate extraction, filtration and clean-up of solid and semisolid sample matrices using common solvents at elevated temperatures and pressures. Low- and high-throughput systems are available for a wide variety of applications.
Dionex ASE systems enable extraction of sample sizes 1 to 100g in minutes, perform unattended extraction of up to 24 samples and use 50 to 90% less solvent compared to other methods. The chemically inert pathway supports acidic and alkaline matrices and solvents. These flexible, easy-to-operate, cost-effective systems have been approved for use by the U.S. EPA and CLP Program.
There are a variety of products that can be added to the Dionex ASE extraction cell to remove moisture, disperse the sample matrix and neutralize mineral acids.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Quantity1
Volume (Metric)200 g
For Use With (Equipment)Dionex ASE Systems
TypeDionex ASE Prep MAP, Moisture Absorbing Polymer
Unit SizeEach 1