Specifications
DescriptionCation-exchange hydronium form
For Use With (Equipment)0 to 100% HPLC Solvents, Dionex OnGuard Sample Prep Station
TypeOnGuard II Ag/H Cartridge, 2.55-cc
pH0 to 14
Quantity12
Sample CapacityFormat not available (1 cc);4.6 (Ag), 0.8 (H) (2.5 cc)
Unit SizeEach 1