Acclaim™ SEC-1000 Size Exclusion Chromatography HPLC Columns
Product Image
Thermo Scientific™

Acclaim™ SEC-1000 Size Exclusion Chromatography HPLC Columns

Separate water-soluble polymers and oligomers with the Acclaim family of resin-based, high-performance, size-exclusion chromatography columns.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
079724300 mm4.6 mm
079721300 mm7.8 mm
079722150 mm7.8 mm
08273933 mm4.6 mm
Catalog number 079724
Price (USD)/ Each
2,165.51
Add to cart
Length (Metric):
300 mm
Diameter (Metric):
4.6 mm
Price (USD)/ Each
2,165.51
Add to cart
Acclaim™ SEC-1000 Size Exclusion Chromatography HPLC Columns
Catalog number079724
Price (USD)/ Each
2,165.51
-
Add to cart

Separate water-soluble polymers and oligomers with Thermo Scientific™ Acclaim™ SEC-1000 Size Exclusion HPLC Columns. These resin-based, high-performance, size-exclusion chromatography columns cover molecular weight range: 7μm, 1000Å. The combination of a proprietary mono-dispersed multi-pore hydrophilic resin and small particle size provides high-resolution and good linearity of the calibration curve with no inflection points for accurate, reliable analysis of a variety of water-soluble polymers.

The Acclaim SEC-1000 LC column is calibrated from 1,000 to 1,000,000 Daltons and can characterize high molecular weight polymers.

High-Performance Analysis of Water-Soluble Polymers
  • High-resolution analysis at reduced solvent consumption in available 4.6 x 300mm dimension
  • Stable surface bonding leads to low column bleed and enhances compatibility with UV, RI, MS, ELSD and Thermo Scientific Dionex™ Corona™ Charged Aerosol Detectors
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Flow Rate0.35 mL/min
Length (Metric)300 mm
Max. Pressure600 psi (40 bar)
Particle ShapeSpherical
Particle Size7 μm
pH2 to 12
Pore Size1000 Å
Temperature60°C
Stationary PhaseProprietary
Product LineAcclaim
Packing MaterialSpherical, Fully Porous Polymer
Column TypeSize Exclusion
EndcappedYes
PhaseSize Exclusion
TypeAnalytical Column
TechniqueBio-LC
Unit SizeEach