Dionex™ ICS-6000 AM Automation Manager
Dionex™ ICS-6000 AM Automation Manager
Actual product may vary
Thermo Scientific™
Dionex™ ICS-6000 AM Automation Manager
Simplify and automate ion chromatography sample preparation, preconcentration, matrix elimination, and postcolumn reagent additions in Dionex ICS-6000 HPIC systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
075950Automation Manager with two 10-port High-Pressure Valves and two Low-Pressure 3-Way Valves
079833Automation Manager (no valves installed)
075951Automation Manager with one 10-port High Pressure Valve and one Low Pressure 3-Way Valve
075952Automation Manager with one 6-port High Pressure Valve and one Low Pressure 3-Way Valve
Catalog number 075950
Price (USD)/ Each
-
Request A Quote
Description:
Automation Manager with two 10-port High-Pressure Valves and two Low-Pressure 3-Way Valves
Save time and streamline even complex ion chromatography (IC) applications, including automated sample preparation, preconcentration, matrix elimination, and postcolumn reagent addition. Configure the Thermo Scientific™ Dionex™ ICS-6000 AM Automation Manager in the upper compartment of the Dionex DC Detector/Chromatography Compartment with a choice of two high-pressure rotary valves or two low-pressure solenoid valves. Setup is easy with components recognized, configured and controlled through Thermo Scientific™ Chromeleon™ Chromatography Data System software.
  • Complete automation of even complex applications
  • Full support for standard bore and microbore chemistries on the Dionex™ ICS-6000 HPIC™ Standard and Microbore and Hybrid systems
  • Ability to streamline postcolumn reagent addition using Thermo Scientific™ Dionex™ AutoPrep™ and RFIC™-ESP options
Not available for capillary IC.
Specifications
For Use With (Equipment)Dionex ICS-5000+/ICS-6000 Ion Chromatography Systems with Chromeleon Software
TypeAutomation Manager
OperationStreamlines and automates ion chromatography (IC) applications.
DescriptionAutomation Manager with two 10-port High-Pressure Valves and two Low-Pressure 3-Way Valves
Unit SizeEach