Be ready for any ion chromatography (IC) application, from single column methods in high-background matrices to trace contaminant determinations in high-purity water. The Thermo Scientific™ Dionex™ ICS-4000 CD Conductivity Detector handles even very high or low concentrations with high sensitivity, low noise and without cumbersome range change distortions.