Dionex™ ICS-5000+/ICS-6000 ED Electrochemical Conventional Electrodes
Dionex&trade; ICS-5000<sup>+</sup>/ICS-6000 ED Electrochemical Conventional Electrodes
Actual product may vary
Thermo Scientific™
Dionex™ ICS-5000+/ICS-6000 ED Electrochemical Conventional Electrodes
Obtain Gold, AAA Gold, Platinum, Glassy Carbon or Silver ED electrodes with gasket and polishing kit for Dionex ICS-5000+/ICS-6000 ED Electrochemical Conventional Electrodes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
063722AAA Gold Electrode with Gasket and Polishing Kit
079850Gold Electrode with Gasket and Polishing Kit
079851Platinum Electrode with Gasket and Polishing Kit
079854Glassy Carbon Electrode with Gasket and Polishing Kit
079856Silver Electrode with Gasket and Polishing Kit
0637233 mm Gold Working Electrode with Gasket and Polishing Kit
Catalog number 063722
Price (USD)/ Each 1
1,370.00
Add to cart
Description:
AAA Gold Electrode with Gasket and Polishing Kit
Price (USD)/ Each 1
1,370.00
Add to cart
Dionex™ ICS-5000+/ICS-6000 ED Electrochemical Conventional Electrodes
Catalog number063722
Price (USD)/ Each 1
1,370.00
-
Add to cart
Offer a choice of Gold, AAA Gold, Platinum, Glassy Carbon and Silver, each with gasket and polishing kit with Thermo Scientific™ Dionex™ ICS-5000+/ICS-6000 ED Electrochemical Conventional Electrodes.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)ICS-5000+/ICS-6000 Ion Chromatography Systems
TypeGold Electrode
MaterialAAA Gold
IncludesGasket, polishing kit
DescriptionAAA Gold Electrode with Gasket and Polishing Kit
Unit SizeEach 1