Obtain results you can always count on for the crucial monitoring of everolimus drug levels in kidney and liver transplant patients with the Thermo Scientific™ QMS™ Everolimus Immunoassay. Using our proprietary QMS technology, these liquid, ready-to-use reagents provide excellent accuracy and precision. Applications are available on our compact Indiko™ and Indiko Plus clinical and specialty chemistry systems as well as a wide variety of other laboratory instrumentation.
Key Features- Optimal low-end sensitivity with a broad dynamic range
- Excellent correlation with HPLC-MS, the gold standard method
- Precise throughout the standard assay range
For In Vitro Diagnostic Use.
For In Vitro Diagnostic Use.