You need to enable JavaScript to run this app.
QMS™ Therapeutic Drug Monitoring (TDM) Controls
Actual product may vary
Thermo Scientific™
QMS™ Therapeutic Drug Monitoring (TDM) Controls
Ensure accurate results when testing with the newest TDM reagents with Thermo Scientific™ QMS™ TDM Controls, offered in three levels.
Have Questions?
Change view
Catalog Number
Description
0373373
Zonisamide Controls
0374090
Lamotrigine Controls
0374181
Topiramate Controls
3 Options
Catalog number
0373373
Price (USD)
/ Each
-
Request A Quote
Description:
Zonisamide Controls
Ensure accurate results when testing with the newest TDM reagents with Thermo Scientific™ QMS™ TDM Controls, offered in three levels.
For In Vitro Diagnostic Use.
For In Vitro Diagnostic Use.
Specifications
Description
Zonisamide Controls
Concentration
8.0, 25.0, 50.0 μg/mL
Detectable Analytes
Zonisamide
Sample Type
Plasma, Serum
Type
Control
Level
Level 3
Quantity
3 x 2.5 mL
Unit Size
Each
Contents & Storage
2°C to 8°C