DRI™ Phencyclidine (PCP) Drugs of Abuse Assays
Product Image
Thermo Scientific™

DRI™ Phencyclidine (PCP) Drugs of Abuse Assays

Eliminate time-consuming reagent preparation yet deliver high analytical accuracy with Thermo Scientific™ DRI™ Phencyclidine (PCP) Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
01601 x 100 mL
01611 x 500 mL
100146733 x 18 mL
Catalog number 0160
Price (USD)/ Each
-
Request A Quote
Quantity:
1 x 100 mL
Eliminate time-consuming reagent preparation yet deliver high analytical accuracy with Thermo Scientific™ DRI™ Phencyclidine (PCP) Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. These liquid, ready-to-use assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionDRI Phencyclidine (PCP) Drugs of Abuse Assays
Detectable AnalytesPhencyclidine (PCP)
DoA CalibratorsDRI Multi-drug Calibrators
Sample TypeUrine
TypeReagent
Quantity1 x 100 mL
Unit SizeEach
Contents & Storage
2°C to 8°C