Orion™ 2 Cell Conductivity Probes
Orion™ 2 Cell Conductivity Probes
Actual product may vary
Thermo Scientific™
Orion™ 2 Cell Conductivity Probes
Perform applications in either the lab or the field with these versatile 2-electrode conductivity cells, available in a variety of materials and measurement ranges.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMeasurement RangesConnector TypesFor Use With (Equipment)
013016MD0.01 to 300 μS/cmMini-DINOrion Star Series Meters
011050MD1 μS/cm to 20 mS/cmMini-DINOrion Star Series Meters
011510MD10 μS/cm to 200 mS/cm8-pin Mini-DINOrion Star Series Meters
018020MD10 μS/cm to 2000 mS/cmMini-DINOrion Star Series Meters
Catalog number 013016MD
Price (USD)/ Each
1,010.00
-
Add to cart
Measurement Ranges:
0.01 to 300 μS/cm
Connector Types:
Mini-DIN
For Use With (Equipment):
Orion Star Series Meters
Price (USD)/ Each
1,010.00
Add to cart
Orion™ 2 Cell Conductivity Probes
Catalog number013016MD
Price (USD)/ Each
1,010.00
-
Add to cart
Achieve accurate measurements from ultrapure water, sea water, and more with Thermo Scientific™ Orion™ 2-Electrode Cells. These probes use two electrode cells to measure low-, standard-, and high-conductivity samples based on the cell constant. They are a versatile option for precise measurements in the lab and field. 013016MD is ideal in low ionic strength solutions, such as deionized water and ultrapure water when paired with a flow-cell assembly. 018020MD is good for chemically reactive conductivity samples, including concentrated acids and bases. 011050MD and 011510MD epoxy/graphite cells are durable, chemical resistant, and measure a wide range of conductivities in the lab or field.
The ThermoFisher Orion Conductivity electrode 013016MD complies with SM 2510B; D1125-95, -99 (A) for Drinking water measurements and  USP<645> Measuring the Conductivity of Pure Water
Features
These probes use two electrode cells to measure low-, standard-, and high-conductivity samples based on the cell constant. They are a versatile option for precise measurements in the lab and field.
013016MD is ideal in low ionic strength solutions, such as deionized water and ultrapure water when paired with a flow-cell assembly . 018020MD is good for chemically reactive conductivity samples, including concentrated acids and bases.  011050MD and 011510MD epoxy/graphite cells are durable, chemical resistant, and measure a wide range of conductivities in the lab or field.
Solutions required
Standards
011008               100 µS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100910           147 µS/cm conductivity standard, 10 pouches
011007               1413 µS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100710           1413 µS/cm conductivity/TDS standard, 10 pouches
011006               12.9 mS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100610           12.9 mS/cm conductivity/TDS standard, 10 pouches
011005               111 mS/cm conductivity standard, 5 x 60 mL bottles
01100510           111 mS/cm conductivity standard, 10 pouches
Conditioning solution
011001 Conditioning solution for 011050MD conductivity cells
Accessories
013017               Replacement flow cell
Items included
011050MD         011007 1413 µS/cm conductivity/TDS standard, 60 mL 1 bottle
                          011001 Conditioning solution, 60 mL 1 bottle
011510MD         011006 12.9 mS/cm conductivity/TDS standard, 60 mL 1 bottle
                          011007 1413 µS/cm conductivity/TDS standard, 60 mL 1 bottle
013016MD         011008 100 µS/cm conductivity/TDS standard, 60 mL 1 bottle
                           013017 Replacement flow cell
Warranty period
12 months from date of purchase
Specifications
For Use With (Equipment)Orion Star Series Meters
Cell Constant0.1 cm^-1
Type2-Electrode Conductivity Cells
Connector TypesMini-DIN
DescriptionOrion 2 Cell Ultra-pure Conductivity Probe
MaterialSteel, V4A
Measurement Ranges0.01 to 300 μS/cm
Unit SizeEach