CAS-Block is a powerful universal blocking agent for immunochemistry methods including immunoperoxidase, HistoGold, Western blotting, etc. It has proven superiorRead more
Have Questions?
Catalog number 008120
Price (USD)/ 100 mL
140.65
Online exclusive
149.00
Save 8.35 (6%)
-
Add to cart
Price (USD)/ 100 mL
140.65
Online exclusive
149.00
Save 8.35 (6%)
Add to cart
CAS-Block™ Histochemical Reagent
Catalog number008120
Price (USD)/ 100 mL
140.65
Online exclusive
149.00
Save 8.35 (6%)
-
Add to cart
CAS-Block is a powerful universal blocking agent for immunochemistry methods including immunoperoxidase, HistoGold, Western blotting, etc. It has proven superior to traditional blocking methods and eliminates the need for species-specific serum blockers.