Histomount Mounting Solution
Product Image
Invitrogen™
Histomount Mounting Solution
This mounting solution is designed for permanent mounting of DAB and BCIP/NBT. Dehydrate tissue before use. This is the sameRead more
Have Questions?
Catalog number 008030
Price (USD)/ 15 ml
64.65
Online exclusive
67.00 
Save 2.35 (4%)
-
Add to cart
Price (USD)/ 15 ml
64.65
Online exclusive
67.00 
Save 2.35 (4%)
Add to cart
Histomount Mounting Solution
Catalog number008030
Price (USD)/ 15 ml
64.65
Online exclusive
67.00 
Save 2.35 (4%)
-
Add to cart
This mounting solution is designed for permanent mounting of DAB and BCIP/NBT. Dehydrate tissue before use. This is the same solution that is provided in the Histostain® -SP DAB Kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineHistoMount™
Quantity15 mL
Reagent TypeMounting Solution
Shipping ConditionRoom Temperature
Unit Size15 ml
Contents & Storage
Store at room temperature.
Shelf life: 18 months from date of manufacture