eBioscience™ ELISA Wash Buffer, 1L packets
Product Image
Invitrogen™
eBioscience™ ELISA Wash Buffer, 1L packets
The eBioscience ELISA Wash Buffer Powder is a dried powder formulation of phosphate buffered saline with 0.05% Tween-20. Each packetRead more
Have Questions?
Catalog number 00-0400-46
Price (USD)/ 10 Packets
172.00
-
Add to cart
Price (USD)/ 10 Packets
172.00
Add to cart
eBioscience™ ELISA Wash Buffer, 1L packets
Catalog number00-0400-46
Price (USD)/ 10 Packets
172.00
-
Add to cart
The eBioscience ELISA Wash Buffer Powder is a dried powder formulation of phosphate buffered saline with 0.05% Tween-20. Each packet is reconstituted in 1 Liter of distilled, deionized water; the resulting 1x PBS/Tween-20 buffer (pH 7.4) is suitable for washing ELISA plate microwells. 10 packets of ELISA Wash Buffer Powder are supplied, yielding a total of 10 liters of 1x Wash Buffer.
Reported Application
ELISA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeWash Buffer
FormPowder
For Use With (Application)ELISA
Product LineeBioscience™
Unit Size10 Packets