Live HeLa cells stained with the reagents in the Image-iT® LIVE Plasma Membrane and Nuclear Labeling Kit.
Live, highly confluent HeLa cells were transfected using pShooter vector pCMV/myc/mito/GFP and Lipofectamine 2000 transfection reagent and stained with the reagents in the Image-iT® LIVE Plasma Membrane and Nuclear Labeling Kit (I34406). Plasma membranes were labeled with Alexa Fluor® 594 wheat germ agglutinin and nuclei were stained with Hoechst 33342. Cells were visualized using epifluorescence microscopy.
Related Products
Related Images
Live cell imaging with CellLight™ reagents. Go ›
Imaging autophagy in live HeLa cells with CellLight® reagents for mitochondria and lysosomes: Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›