Rat fibroblasts. LysoTracker® Yellow HCK-123 and LysoTracker® Red DND-99.
Rat fibroblasts stained with LysoTracker® Yellow HCK-123 (L12491) and LysoTracker® Red DND-99 (L7528). The lysosomes fluoresce green when visualized through a bandpass filter set appropriate for fluorescein (bottom image) and red when visualized through a longpass filter set appropriate for rhodamine (top image).