HL-60 cells. ChromaTide BODIPY FL-14-dUTP.
HL-60 cells treated with camptothecin for three hours. The DNA strand nicks characteristic of apoptosis were detected with the TUNEL (terminal deoxynucleotidyl transferase–mediated dUTP nick end-labeling) assay using the fluorescently labeled nucleotide ChromaTide BODIPY FL-14-dUTP (C7614). Image contributed by Zbigniew Darzynkiewicz, Cancer Research Institute, New York Medical College.
Related Products
Related Images
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Agarose gel containing camptothecin-treated HL-60 cells. Go ›
Imaging autophagy in live HeLa cells with CellLight® reagents for mitochondria and lysosomes: Go ›